Ethanolamine hydrochloride, 99+%
Ethanolamine hydrochloride, 99+%
Ethanolamine hydrochloride, 99+%
Ethanolamine hydrochloride, 99+%
Thermo Scientific Chemicals
Ethanolamine hydrochloride, 99+%
CAS: 2002-24-6 | C2H7NO·HCl | 97.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number FSA217042500
Price (MYR)
626.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
626.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthanolamine hydrochloride
Name Notep.a.
CAS2002-24-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: