Butyl methacrylate, 99%, stabilized
Butyl methacrylate, 99%, stabilized
Butyl methacrylate, 99%, stabilized
Butyl methacrylate, 99%, stabilized
Thermo Scientific Chemicals
Butyl methacrylate, 99%, stabilized
CAS: 97-88-1 | C8H14O2 | 142.20 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA2166200101 LGlass bottle
Catalog number FSA216620010
Price (MYR)
403.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
403.00
EA
Add to cart
Specifications
Chemical Name or MaterialButyl methacrylate
Name NoteStabilized
CAS97-88-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapor.