3-Glycidoxypropyltrimethoxysilane, 97%
3-Glycidoxypropyltrimethoxysilane, 97%
3-Glycidoxypropyltrimethoxysilane, 97%
3-Glycidoxypropyltrimethoxysilane, 97%
Thermo Scientific Chemicals
3-Glycidoxypropyltrimethoxysilane, 97%
CAS: 2530-83-8 | C9H20O5Si | 236.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA216545000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Glycidoxypropyltrimethoxysilane
CAS2530-83-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye damage.
Health Hazard 3GHS P Statement
Wear face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.