1,1,3,3-Tetramethyldisiloxane, 97%
1,1,3,3-Tetramethyldisiloxane, 97%
1,1,3,3-Tetramethyldisiloxane, 97%
1,1,3,3-Tetramethyldisiloxane, 97%
Thermo Scientific Chemicals
1,1,3,3-Tetramethyldisiloxane, 97%
CAS: 3277-26-7 | C4H14OSi2 | 134.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA216511000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point70.0°C to 71.0°C
Chemical Name or Material1,1,3,3-Tetramethyldisiloxane
Melting Point-78.0°C
CAS3277-26-7
ColorColorless
View more
Reduction reagent
RUO – Research Use Only