Octamethylcyclotetrasiloxane, 98%
Octamethylcyclotetrasiloxane, 98%
Octamethylcyclotetrasiloxane, 98%
Octamethylcyclotetrasiloxane, 98%
Thermo Scientific Chemicals
Octamethylcyclotetrasiloxane, 98%
CAS: 556-67-2 | C8H24O4Si4 | 296.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA216475000
Price (MYR)
2,401.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,401.00
EA
Add to cart
Specifications
Chemical Name or MaterialOctamethylcyclotetrasiloxane
CAS556-67-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0