1,4-Dichlorobutane, 99%
1,4-Dichlorobutane, 99%
1,4-Dichlorobutane, 99%
1,4-Dichlorobutane, 99%
Thermo Scientific Chemicals
1,4-Dichlorobutane, 99%
CAS: 110-56-5 | C4H8Cl2 | 127.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number FSA216435000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 4-Dichlorobutane
Name Note99+%
CAS110-56-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning