Butyl acrylate, 99+%, stabilized
Butyl acrylate, 99+%, stabilized
Butyl acrylate, 99+%, stabilized
Butyl acrylate, 99+%, stabilized
Thermo Scientific Chemicals
Butyl acrylate, 99+%, stabilized
CAS: 141-32-2 | C7H12O2 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Plastic drum
Catalog number FSA215860100
Price (MYR)
1,440.00
EA
Add to cart
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
1,440.00
EA
Add to cart
Specifications
Chemical Name or MaterialButyl acrylate
Acidity0.01% max. (as acrylic acid)
CAS141-32-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful to aquatic li