Ethyl methacrylate, 99%, stabilized
Ethyl methacrylate, 99%, stabilized
Ethyl methacrylate, 99%, stabilized
Ethyl methacrylate, 99%, stabilized
Thermo Scientific Chemicals
Ethyl methacrylate, 99%, stabilized
CAS: 97-63-2 | C6H10O2 | 114.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Catalog number FSA215835000
Price (MYR)
496.00
BOT
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
496.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthyl methacrylate
Name Note99%
CAS97-63-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
Highly flammable liquid and vapor.