Molybdenum(IV) sulfide, 98.5%
Molybdenum(IV) sulfide, 98.5%
Molybdenum(IV) sulfide, 98.5%
Molybdenum(IV) sulfide, 98.5%
Thermo Scientific Chemicals
Molybdenum(IV) sulfide, 98.5%
CAS: 1317-33-5 | MoS2 | 160.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA215785000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMolybdenum(IV) sulfide
Melting Point2375.0°C
CAS1317-33-5
ColorBlack or Gray
Health Hazard 1GHS Signal Word: Warning