Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Thermo Scientific Chemicals
Phenylmethanesulfonyl fluoride, 99%
CAS: 329-98-6 | C7H7FO2S | 174.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 g
5 g
10 g
Packaging:
Glass bottle
Catalog number FSA215740500
Price (MYR)
2,059.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,059.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhenylmethanesulfonyl fluoride
CAS329-98-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0