Tri-n-butyltin hydride, 97%
Tri-n-butyltin hydride, 97%
Tri-n-butyltin hydride, 97%
Tri-n-butyltin hydride, 97%
Thermo Scientific Chemicals
Tri-n-butyltin hydride, 97%
CAS: 688-73-3 | C12H28Sn | 291.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA215732500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTri-n-butyltin hydride
CAS688-73-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes damage to organs through prolonged
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiou
View more
Reduction reagent
RUO – Research Use Only