Calcium phosphate, dibasic, 97%, pure, anhydrous
Calcium phosphate, dibasic, 97%, pure, anhydrous
Calcium phosphate, dibasic, 97%, pure, anhydrous
Calcium phosphate, dibasic, 97%, pure, anhydrous
Thermo Scientific Chemicals
Calcium phosphate, dibasic, 97%, pure, anhydrous
CAS: 7757-93-9 | CaHO4P | 136.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA215710010
Price (MYR)
651.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
651.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCalcium phosphate, dibasic
CAS7757-93-9
ColorWhite
Physical FormPowder
Quantity1 kg