Tetracosanoic acid, 99%
Tetracosanoic acid, 99%
Tetracosanoic acid, 99%
Tetracosanoic acid, 99%
Thermo Scientific Chemicals
Tetracosanoic acid, 99%
CAS: 557-59-5 | C24H48O2 | 368.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
5 g
Packaging:
Glass bottle
Catalog number FSA215642500
Price (MYR)
456.00
EA
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
456.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetracosanoic acid
CAS557-59-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective cloth