Methyl laurate, 98+%
Methyl laurate, 98+%
Methyl laurate, 98+%
Methyl laurate, 98+%
Thermo Scientific Chemicals
Methyl laurate, 98+%
CAS: 111-82-0 | C13H26O2 | 214.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
2.5 g
10 g
Packaging:
Glass bottle
Catalog number FSA215560500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl laurate
CAS111-82-0
Health Hazard 1GHS Signal Word: Warning
Melting Point4°C to 5°C
ColorColorless to Yellow