1-Bromohexadecane, 97%
1-Bromohexadecane, 97%
1-Bromohexadecane, 97%
1-Bromohexadecane, 97%
Thermo Scientific Chemicals
1-Bromohexadecane, 97%
CAS: 112-82-3 | C16H33Br | 305.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA215432500
Price (MYR)
420.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
420.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Bromohexadecane
CAS112-82-3
Melting Point17.3°C
ColorAmber to Black-Brown
Density0.9990g/mL