Alloxan monohydrate, 98%
Alloxan monohydrate, 98%
Alloxan monohydrate, 98%
Alloxan monohydrate, 98%
Thermo Scientific Chemicals
Alloxan monohydrate, 98%
CAS: 3237-50-1 | C4H4N2O5 | 160.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA215370100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAlloxan monohydrate
Name Note98%
CAS3237-50-1
CAS Min %97.5
CAS Max %100.0