Dimidium bromide, 98%
Dimidium bromide, 98%
Dimidium bromide, 98%
Dimidium bromide, 98%
Thermo Scientific Chemicals
Dimidium bromide, 98%
CAS: 518-67-2 | C20H18BrN3 | 380.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number FSA215342500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDimidium bromide
Name Note98%
CAS518-67-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement