1-Hexanethiol, 96%
1-Hexanethiol, 96%
1-Hexanethiol, 96%
Thermo Scientific Chemicals
1-Hexanethiol, 96%
CAS: 111-31-9 | C6H14S | 118.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number FSA215271000
Price (MYR)
601.00
Bottle
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
601.00
Bottle
Specifications
Chemical Name or Material1-Hexanethiol
CAS111-31-9
CAS Min %95.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0