Starch, for biochemistry, potato, hydrolyzed for electrophoresis
Starch, for biochemistry, potato, hydrolyzed for electrophoresis
Starch, for biochemistry, potato, hydrolyzed for electrophoresis
Starch, for biochemistry, potato, hydrolyzed for electrophoresis
Thermo Scientific Chemicals
Starch, for biochemistry, potato, hydrolyzed for electrophoresis
CAS: 9005-25-8 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA215125000
Price (MYR)
1,527.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,527.00
BOT
Add to cart
Specifications
Chemical Name or MaterialStarch
ColorWhite
Name NoteHydrolyzed for Electrophoresis
CAS9005-25-8
Physical FormFine Crystalline Powder