Oleic acid, sodium salt, 65-90% oleic C18
Oleic acid, sodium salt, 65-90% oleic C18
Oleic acid, sodium salt, 65-90% oleic C18
Oleic acid, sodium salt, 65-90% oleic C18
Thermo Scientific Chemicals
Oleic acid, sodium salt, 65-90% oleic C18
CAS: 143-19-1 | C18H33NaO2 | 304.44 g/mol
Have Questions?
Catalog NumberQuantity
FSA2150800101 g
Catalog number FSA215080010
Price (MYR)
432.00
BOT
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
432.00
BOT
Add to cart
Specifications
Chemical Name or MaterialOleic acid, sodium salt
Name Note65-90% oleic C18
CAS143-19-1
Melting Point232°C to 235°C
ColorWhite to Yellow