Kimble™ KIMAX™ Distilling Apparatus Complete Assembly
Kimble™ KIMAX™ Distilling Apparatus Complete Assembly
Actual product may vary
Thermo Scientific Chemicals
Kimble™ KIMAX™ Distilling Apparatus Complete Assembly
Complete assembly
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityStandard Taper Stopper No.For Use With (Application)
FSA21500100033.81 oz., 1000mL27General purpose distillation and phenol distillation
ACR21500100033.81 oz., 1000mL27General purpose distillation and phenol distillation
Catalog number FSA215001000
Price (MYR)
615.00
BOT
Add to cart
Capacity:
33.81 oz., 1000mL
Standard Taper Stopper No.:
27
For Use With (Application):
General purpose distillation and phenol distillation
Price (MYR)
615.00
BOT
Add to cart
Use to distill small amounts of water or in APHA Method 4500-NH3 for determining nitrogen (ammonia) in purified drinking water, natural water and highly purified wastewaters (concentration, <20μg/L)
Specifications
Includes500 or 1000mL flat-bottom flask, distillation head with a 19/38 joint stopper, jacketed graham condenser with 19/38 top joint
For Use With (Application)General purpose distillation and phenol distillation
TypeDistillation Apparatus
MaterialBorosilicate Glass
Capacity33.81 oz., 1000mL
Certifications/ComplianceUSP Type I and ASTM E438, Type I, Class A
Capacity (Metric)1000mL
Standard Taper Stopper No.27
Standard Taper Joint19/38
Capacity (English)33.81 oz.
Unit SizeBOT