4'-(Trifluoromethyl)acetophenone, 99%
4'-(Trifluoromethyl)acetophenone, 99%
4'-(Trifluoromethyl)acetophenone, 99%
4'-(Trifluoromethyl)acetophenone, 99%
Thermo Scientific Chemicals
4'-(Trifluoromethyl)acetophenone, 99%
CAS: 709-63-7 | C9H7F3O | 188.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA214450010
Price (MYR)
262.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
262.00
EA
Add to cart
Specifications
Chemical Name or Material4'-(Trifluoromethyl)acetophenone
CAS709-63-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0