Pyridinium p-toluenesulfonate, 98%
Pyridinium p-toluenesulfonate, 98%
Pyridinium p-toluenesulfonate, 98%
Pyridinium p-toluenesulfonate, 98%
Thermo Scientific Chemicals
Pyridinium p-toluenesulfonate, 98%
CAS: 24057-28-1 | C12H13NO3S | 251.3 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA213740250
Price (MYR)
428.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
428.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPyridinium p-toluenesulfonate
CAS24057-28-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER