Calcium phosphate, for analysis, 35-40% (Ca), mixture of Calcium phosphates
Calcium phosphate, for analysis, 35-40% (Ca), mixture of Calcium phosphates
Calcium phosphate, for analysis, 35-40% (Ca), mixture of Calcium phosphates
Calcium phosphate, for analysis, 35-40% (Ca), mixture of Calcium phosphates
Thermo Scientific Chemicals
Calcium phosphate, for analysis, 35-40% (Ca), mixture of Calcium phosphates
CAS: 7758-87-4 | Ca3O8P2 | 310.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number FSA213480010
Price (MYR)
478.00
BOT
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
478.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCalcium phosphate
Melting Point1670.0°C
Name Notec.p.
CAS7758-87-4
Quantity1 kg