Antimony(III) oxide, 99+%
Antimony(III) oxide, 99+%
Antimony(III) oxide, 99+%
Antimony(III) oxide, 99+%
Antimony(III) oxide, 99+%
Thermo Scientific Chemicals
Antimony(III) oxide, 99+%
CAS: 1309-64-4 | O3Sb2 | 291.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
1 kg
Catalog number FSA213470051
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Request bulk or custom format
Specifications
Boiling Point1550°C
Chemical Name or MaterialAntimony(III) oxide
Melting Point656°C
CAS1309-64-4
Health Hazard 1GHS Signal Word: Warning
View more
Suitable for battery materials development.
RUO – Research Use Only