Nitromethane, 99+%, for analysis
Nitromethane, 99+%, for analysis
Nitromethane, 99+%, for analysis
Nitromethane, 99+%, for analysis
Thermo Scientific Chemicals
Nitromethane, 99+%, for analysis
CAS: 75-52-5 | CH3NO2 | 61.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA213375000
Price (MYR)
749.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
749.00
BOT
Add to cart
Specifications
Chemical Name or MaterialNitromethane
Name Notep.a.
CAS75-52-5
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning
View more
For analysis
RUO – Research Use Only