1-Hexene, 97%
1-Hexene, 97%
1-Hexene, 97%
Thermo Scientific Chemicals
1-Hexene, 97%
1-Hexene, 97%, C6H12, CAS Number-592-41-6, dialene 6, butyl ethylene, 1-hexene, butylethylene, 1-n-hexene, hexylene, hexene-1, hexene, n-hexene, unii-b38zz8c206, 1L, 96% min. (GC), 01, 215, 62.0 deg.C to 65.0 deg.C, 100.0, 96.0, CHEBI:24579, Colorless, 0.6780g/mL, 209-753-1, -26 deg.C, 84.15 | CAS: 592-41-6 | C6H12 | 84.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal Drum
Catalog number FSA213210250
Price (MYR)
3,158.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Price (MYR)
3,158.00
EA
Add to cart
Specifications
Chemical Name or Material1-Hexene
CAS592-41-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0