Copper(I) bromide-dimethyl sulfide complex, 99%
Copper(I) bromide-dimethyl sulfide complex, 99%
Copper(I) bromide-dimethyl sulfide complex, 99%
Copper(I) bromide-dimethyl sulfide complex, 99%
Thermo Scientific Chemicals
Copper(I) bromide-dimethyl sulfide complex, 99%
CAS: 54678-23-8 | C2H6BrCuS | 205.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA213180250
Price (MYR)
1,090.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,090.00
EA
Add to cart
Specifications
Chemical Name or MaterialCopper(I) bromide-dimethyl sulfide complex
Name Note99%
CAS54678-23-8
Health Hazard 1MOISTURE SENSITIVE
Health Hazard 2AIR SENSITIVE