Selenourea, 98+%
Selenourea, 98+%
Selenourea, 98+%
Thermo Scientific Chemicals
Selenourea, 98+%
CAS: 630-10-4 | CH4N2Se | 123.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA213170050
Price (MYR)
2,263.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,263.00
EA
Add to cart
Specifications
Chemical Name or MaterialSelenourea
CAS630-10-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.