Magnesium sulfate heptahydrate, 99.5%, for analysis
Magnesium sulfate heptahydrate, 99.5%, for analysis
Magnesium sulfate heptahydrate, 99.5%, for analysis
Magnesium sulfate heptahydrate, 99.5%, for analysis
Thermo Scientific Chemicals
Magnesium sulfate heptahydrate, 99.5%, for analysis
CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number FSA213115000
Price (MYR)
552.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
552.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMagnesium sulfate heptahydrate
Melting Point770.0°C
Name Notep.a.
CAS10034-99-8