Triethylsilane, 99%
Triethylsilane, 99%
Triethylsilane, 99%
Thermo Scientific Chemicals
Triethylsilane, 99%
CAS: 617-86-7 | C6H16Si | 116.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA212921000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point107.0°C to 108.0°C
Chemical Name or MaterialTriethylsilane
CAS617-86-7
ColorColorless
Health Hazard 1GHS Signal Word: Danger
View more
Reduction reagent
RUO – Research Use Only