Tetrabutylammonium hydroxide, 1M solution in methanol
Tetrabutylammonium hydroxide, 1M solution in methanol
Tetrabutylammonium hydroxide, 1M solution in methanol
Tetrabutylammonium hydroxide, 1M solution in methanol
Thermo Scientific Chemicals
Tetrabutylammonium hydroxide, 1M solution in methanol
CAS: 2052-49-5 | C16H37NO | 259.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
2.5 L
Catalog number FSA212918000
Price (MYR)
3,432.00
BOT
Add to cart
Quantity:
800 mL
Request bulk or custom format
Price (MYR)
3,432.00
BOT
Add to cart
Chemical Identifiers
CAS2052-49-5, 67-56-1
Specifications
Chemical Name or MaterialTetrabutylammonium hydroxide
Name Note1M Solutionin Methanol
CAS2052-49-5
Boiling Point65.0°C
Health Hazard 1Danger
View more
Phase transfer catalyst
RUO – Research Use Only