Potassium antimonyl tartrate trihydrate, ACS reagent
Potassium antimonyl tartrate trihydrate, ACS reagent
Potassium antimonyl tartrate trihydrate, ACS reagent
Potassium antimonyl tartrate trihydrate, ACS reagent
Thermo Scientific Chemicals
Potassium antimonyl tartrate trihydrate, ACS reagent
CAS: 28300-74-5 | 1·5 H2O | 333.93 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA212801000100 gGlass bottle
Catalog number FSA212801000
Price (MYR)
676.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
676.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium antimonyl tartrate sesquihydrate
CAS28300-74-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.