Iodine, 99.5+%, for analysis
Iodine, 99.5+%, for analysis
Iodine, 99.5+%, for analysis
Iodine, 99.5+%, for analysis
Thermo Scientific Chemicals
Iodine, 99.5+%, for analysis
C-X bond formation reagent | CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA212491000
Price (MYR)
543.00
BOT
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
543.00
BOT
Add to cart
Specifications
Boiling Point185.0°C
Chemical Name or MaterialIodine
Melting Point113.0°C
CAS7553-56-2
Health Hazard 1Danger