Cadmium acetate dihydrate, 98%, pure
Cadmium acetate dihydrate, 98%, pure
Cadmium acetate dihydrate, 98%, pure
Cadmium acetate dihydrate, 98%, pure
Thermo Scientific Chemicals
Cadmium acetate dihydrate, 98%, pure
CAS: 5743-04-4 | C4H6CdO4·2H2O | 266.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number FSA212310010
Price (MYR)
1,629.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
1,629.00
EA
Add to cart
Specifications
Chemical Name or MaterialCadmium acetate dihydrate
CAS5743-04-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Very toxic to aquatic life with long lasting effects.
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a posi