Aluminum oxide, 99.99%, (trace metal basis), extra pure
Aluminum oxide, 99.99%, (trace metal basis), extra pure
Aluminum oxide, 99.99%, (trace metal basis), extra pure
Aluminum oxide, 99.99%, (trace metal basis), extra pure
Thermo Scientific Chemicals
Aluminum oxide, 99.99%, (trace metal basis), extra pure
CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA212251000
Price (MYR)
1,151.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,151.00
EA
Add to cart
Specifications
Boiling Point2977.0°C
Chemical Name or MaterialAluminum oxide
Melting Point2030.0°C
Name NoteExtra Pure
CAS1344-28-1
View more
For ultra trace analysis
RUO – Research Use Only