Nuclear Fast Red, 94%, pure
Nuclear Fast Red, 94%, pure
Nuclear Fast Red, 94%, pure
Nuclear Fast Red, 94%, pure
Thermo Scientific Chemicals
Nuclear Fast Red, 94%, pure
CAS: 6409-77-4 | C14H11NNaO7S | 360.292 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA211980050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialNuclear Fast Red
Melting Point>300.0°C
CAS6409-77-4
CAS Min %92.0
CAS Max %100.0