Benzyltrimethylammonium chloride, 98+%
Benzyltrimethylammonium chloride, 98+%
Benzyltrimethylammonium chloride, 98+%
Benzyltrimethylammonium chloride, 98+%
Thermo Scientific Chemicals
Benzyltrimethylammonium chloride, 98+%
CAS: 56-93-9 | C10H16ClN | 185.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Glass bottle
Catalog number FSA211862500
Price (MYR)
422.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
422.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzyltrimethylammonium chloride
Name Note98+%
CAS56-93-9
CAS Min %98.0
Health Hazard 1GHS Signal Word: Danger
View more
Phase transfer catalyst
RUO – Research Use Only