Cerium(III) chloride heptahydrate, 99%
Cerium(III) chloride heptahydrate, 99%
Cerium(III) chloride heptahydrate, 99%
Cerium(III) chloride heptahydrate, 99%
Thermo Scientific Chemicals
Cerium(III) chloride heptahydrate, 99%
CAS: 18618-55-8 | CeCl3·7H2O | 372.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA211825000
Price (MYR)
1,832.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,832.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCerium(III) chloride heptahydrate
Melting Point848.0°C
Name Note99%
CAS18618-55-8
ColorYellow