Dibenzoyl peroxide, 75%, remainder water
Dibenzoyl peroxide, 75%, remainder water
Dibenzoyl peroxide, 75%, remainder water
Dibenzoyl peroxide, 75%, remainder water
Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water
CAS: 94-36-0 | C14H10O4 | 242.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA211781000
Price (MYR)
306.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
306.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDibenzoyl peroxide
Name Note75%, remainder water
CAS94-36-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Causes serious eye irritation.
Heating may cause a fire.
Very toxic to aquatic life with long lasting effects.
View more
Radical initiators
RUO – Research Use Only