Isonicotinoyl chloride hydrochloride, 97%
Isonicotinoyl chloride hydrochloride, 97%
Isonicotinoyl chloride hydrochloride, 97%
Isonicotinoyl chloride hydrochloride, 97%
Thermo Scientific Chemicals
Isonicotinoyl chloride hydrochloride, 97%
CAS: 39178-35-3 | C6H5Cl2NO | 178.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA211680250
Price (MYR)
474.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
474.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIsonicotinoyl chloride hydrochloride
CAS39178-35-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi