Neodymium(III)-oxide, 99.9%, (trace metal basis)
Neodymium(III)-oxide, 99.9%, (trace metal basis)
Neodymium(III)-oxide, 99.9%, (trace metal basis)
Neodymium(III)-oxide, 99.9%, (trace metal basis)
Thermo Scientific Chemicals
Neodymium(III)-oxide, 99.9%, (trace metal basis)
CAS: 1313-97-9 | Nd2O3 | 336.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA211581000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialNeodymium(III)-oxide
Melting Point2270.0°C
Name Note99.9%
CAS1313-97-9
ColorBlue
View more
for ultra trace analysis
RUO – Research Use Only