Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline
Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline
Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline
Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline
Thermo Scientific Chemicals
Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline
CAS: 16674-78-5 | C4H14MgO8 | 214.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number FSA211572500
Price (MYR)
320.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
320.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMagnesium acetate tetrahydrate
Melting Point80.0°C
Name NoteExtra Pure
CAS16674-78-5