Acriflavine neutral, 13.3-15.8% chlorine
Acriflavine neutral, 13.3-15.8% chlorine
Acriflavine neutral, 13.3-15.8% chlorine
Acriflavine neutral, 13.3-15.8% chlorine
Thermo Scientific Chemicals
Acriflavine neutral, 13.3-15.8% chlorine
CAS: 8048-52-0 | C27H25ClN6 | 468.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA211401000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcriflavine neutral
CAS8048-52-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici