Xylenol Orange, sodium salt, pure, water soluble
Xylenol Orange, sodium salt, pure, water soluble
Xylenol Orange, sodium salt, pure, water soluble
Xylenol Orange, sodium salt, pure, water soluble
Thermo Scientific Chemicals
Xylenol Orange, sodium salt, pure, water soluble
CAS: 3618-43-7 | C31H28N2Na4O13S
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 g
5 g
10 g
Packaging:
Glass bottle
Catalog number FSA211220500
Price (MYR)
3,875.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,875.00
EA
Add to cart
Specifications
Chemical Name or MaterialXylenol Orange, sodium salt
Melting Point210.0°C
CAS3618-43-7
Molecular FormulaC31H28N2Na4O13S
Solubility InformationSolubility in water: 510g/L (20°C).
View more
Indicator
RUO – Research Use Only