D-Gluconic acid, calcium salt, 99%
D-Gluconic acid, calcium salt, 99%
D-Gluconic acid, calcium salt, 99%
D-Gluconic acid, calcium salt, 99%
Thermo Scientific Chemicals
D-Gluconic acid, calcium salt, 99%
CAS: 299-28-5 | C12H22CaO14 | 430.372 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA211062500
Price (MYR)
283.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
283.00
BOT
Add to cart
Specifications
Chemical Name or MaterialD-Gluconic acid, calcium salt
CAS299-28-5
ColorWhite
Loss on Drying2.0 % max. (1 g, 105°C, 16 hrs)
Infrared SpectrumAuthentic