Calcium acetate hydrate, 99%, extra pure
Calcium acetate hydrate, 99%, extra pure
Calcium acetate hydrate, 99%, extra pure
Calcium acetate hydrate, 99%, extra pure
Calcium acetate hydrate, 99%, extra pure
Thermo Scientific Chemicals
Calcium acetate hydrate, 99%, extra pure
CAS: 114460-21-8 | C4H6CaO4 | 158.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number FSA211052500
Price (MYR)
407.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
407.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCalcium acetate hydrate
CAS114460-21-8
ColorWhite
pH7.0 to 9.0 (10% soln., 20°C)
Physical FormPowder and/or Agglomerates