Lawesson's Reagent, 99%
Lawesson's Reagent, 99%
Lawesson's Reagent, 99%
Lawesson's Reagent, 99%
Thermo Scientific Chemicals
Lawesson's Reagent, 99%
CAS: 19172-47-5 | C14H14O2P2S4 | 404.452 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA210891000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialLawesson's Reagent
ColorWhite
CAS19172-47-5
Melting Point228.0°C to 231.0°C
Assay Percent Range99%
View more
For C-S bond formation
RUO – Research Use Only