2,2-Dimethyl-1,3-propanediamine, 99%
2,2-Dimethyl-1,3-propanediamine, 99%
2,2-Dimethyl-1,3-propanediamine, 99%
2,2-Dimethyl-1,3-propanediamine, 99%
Thermo Scientific Chemicals
2,2-Dimethyl-1,3-propanediamine, 99%
CAS: 7328-91-8 | C5H14N2 | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA210510500
Price (MYR)
582.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
582.00
EA
Add to cart
Specifications
Chemical Name or Material2, 2-Dimethyl-1, 3-propanediamine
CAS7328-91-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0