Methoxylamine hydrochloride, 98+%
Methoxylamine hydrochloride, 98+%
Methoxylamine hydrochloride, 98+%
Methoxylamine hydrochloride, 98+%
Thermo Scientific Chemicals
Methoxylamine hydrochloride, 98+%
CAS: 593-56-6 | CH5NO·HCl | 83.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA210490250
Price (MYR)
1,097.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,097.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethoxylamine hydrochloride
CAS593-56-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
View more
Alkylation reagent
RUO – Research Use Only